missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10285574
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10285574 1 g 1g
10634785 250 mg 250mg
10443204 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10285574 Supplier Thermo Scientific Maybridge Supplier No. CC56001DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 160388-53-4
Molecular Formula C11H10N2O2
Molecular Weight (g/mol) 202.213
MDL Number MFCD07186451
InChI Key ZHQQRHUITAFMTC-UHFFFAOYSA-N
PubChem CID 6484268
IUPAC Name 4-(pyrazol-1-ylmethyl)benzoic acid
SMILES C1=CN(N=C1)CC2=CC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-1h-pyrazol-1-ylmethyl benzoic acid, 4-pyrazol-1-ylmethyl-benzoic acid, 4-pyrazol-1-ylmethyl benzoic acid, benzoicacid, 4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl benzoic acid, 4-1h-pyrazol-1-yl methyl benzoic acid, 4-1h-pyrazole-1-ylmethyl benzoic acid, benzoicacid,4-1h-pyrazol-1-ylmethyl, benzoic acid, 4-1h-pyrazol-1-ylmethyl
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.