missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10110972
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 17635-45-9
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD00626298
InChI Key CSFIQHZIFKIQNO-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl aniline, 4-pyrazol-1-yl-phenylamine, 4-1-pyrazolyl aniline, 1-4-aminophenyl pyrazole, 4-pyrazol-1-yl aniline, 4-1h-pyrazol-1-yl benzenamine, benzenamine, 4-1h-pyrazol-1-yl, 4-1h-pyrazol-1-yl-phenylamine, 4-pyrazolylphenylamine, 1-p-aminophenylpyrazole
PubChem CID 594274
IUPAC Name 4-pyrazol-1-ylaniline
SMILES C1=CN(N=C1)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'