missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-yl)aniline, 97%, Thermo Scientific™

Product Code. 10019121 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10019121 10 g Amber Glass Bottle 10g
10110972 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10019121 Supplier Thermo Scientific Supplier No. CC18414EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17635-45-9
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD00626298
InChI Key CSFIQHZIFKIQNO-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl aniline, 4-pyrazol-1-yl-phenylamine, 4-1-pyrazolyl aniline, 1-4-aminophenyl pyrazole, 4-pyrazol-1-yl aniline, 4-1h-pyrazol-1-yl benzenamine, benzenamine, 4-1h-pyrazol-1-yl, 4-1h-pyrazol-1-yl-phenylamine, 4-pyrazolylphenylamine, 1-p-aminophenylpyrazole
PubChem CID 594274
IUPAC Name 4-pyrazol-1-ylaniline
SMILES C1=CN(N=C1)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 159.19
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.