missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-imidazol-1-ylmethyl)benzoic acid hydrochloride, Thermo Scientific™

Product Code. 10245754
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245754 250 mg Amber Glass Bottle 250mg
10183464 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245754 Supplier Thermo Scientific Maybridge Supplier No. CC43351CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 135611-32-4
Molecular Formula C11H11ClN2O2
Molecular Weight (g/mol) 238.671
MDL Number MFCD08271922
InChI Key MZTHHEXWAYFTBJ-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-ylmethyl benzoic acid hydrochloride, 4-1h-imidazol-1-yl methyl benzoic acid hydrochloride, 4-imidazol-1-ylmethyl benzoic acid hydrochloride, acmc-1bx8o, 4-imidazolylmethyl benzoic acid, chloride, 4-1h-imidazol-1-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 18525764
IUPAC Name 4-(imidazol-1-ylmethyl)benzoic acid;hydrochloride
SMILES C1=CC(=CC=C1CN2C=CN=C2)C(=O)O.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 238.67
Percent Purity 95%
Chemical Name or Material 4-(1H-imidazol-1-ylmethyl)benzoic acid hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.