missing translation for 'onlineSavingsMsg'
Learn More

4-(1h-imidazol-1-ylmethyl)aniline, Thermo Scientific™

Catalog Number 10236273
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
56643-85-7
C10H11N3
173.22
MFCD06660785
DGHAOTHIDTUSJY-UHFFFAOYSA-N
4-imidazol-1-ylmethylphenylamine, 4-1h-imidazol-1-ylmethyl aniline, 4-imidazol-1-ylmethyl aniline, 4-1h-imidazol-1-yl methyl aniline, 4-1-imidazolyl methyl aniline, 4-imidazol-1-ylmethyl-phenylamine, 1-4-aminobenzyl imidazole, 4-imidazolylmethyl phenylamine, 1-4-aminobenzyl-1h-imidazole
1265319
4-(imidazol-1-ylmethyl)aniline
NC1=CC=C(CN2C=CN=C2)C=C1
This item is not returnable. View return policy
56643-85-7
C10H11N3
173.22
MFCD06660785
DGHAOTHIDTUSJY-UHFFFAOYSA-N
4-imidazol-1-ylmethylphenylamine, 4-1h-imidazol-1-ylmethyl aniline, 4-imidazol-1-ylmethyl aniline, 4-1h-imidazol-1-yl methyl aniline, 4-1-imidazolyl methyl aniline, 4-imidazol-1-ylmethyl-phenylamine, 1-4-aminobenzyl imidazole, 4-imidazolylmethyl phenylamine, 1-4-aminobenzyl-1h-imidazole
1265319
4-(imidazol-1-ylmethyl)aniline
NC1=CC=C(CN2C=CN=C2)C=C1
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'