missing translation for 'onlineSavingsMsg'
Learn More

4-(Imidazol-1-yl)phenol, 97%, Acros Organics™

Product Code. 10605062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10605062 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10605062 Supplier Thermo Scientific Acros Supplier No. 179790050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10041-02-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 204.0°C to 207.0°C
Color Beige-Brown to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C9H8N2O
MDL Number MFCD00005281
Quantity 5 g
Synonym 4-imidazol-1-yl phenol, 4-1h-imidazol-1-yl phenol, 4-imidazol-1-yl-phenol, 4-1-imidazolyl phenol, 1-p-hydroxyphenyl imidazole, phenol, 4-1h-imidazol-1-yl, 1-4-hydroxyphenyl imidazole, 4-imidazolylphenol, 4‐ 1h‐imidazol‐1‐yl phenol compound 1, p-1-imidazolyl phenol
InChI Key CYKCUAPYWQDIKR-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)N1C=CN=C1
IUPAC Name 4-(1H-imidazol-1-yl)phenol
Molecular Weight (g/mol) 160.18
PubChem CID 82315
Formula Weight 160.18
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-(Imidazol-1-yl)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Imidazol-1-yl)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.