missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Benzimidazol-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10049381
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10049381 10 g 10g
10334342 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10049381 Supplier Thermo Scientific Maybridge Supplier No. KM10745EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 139742-50-0
Molecular Formula C15H12N2O2
Molecular Weight (g/mol) 252.273
MDL Number MFCD02180372
InChI Key KLIANJXCGMBICV-UHFFFAOYSA-N
Synonym 4-1h-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzo d imidazol-1-yl methyl benzoic acid, 4-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-yl methyl benzoic acid, maybridge1_005258, acmc-20e4o9, 4-benzimidazolylmethyl benzoic acid, 4-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzimidazole-1-ylmethyl benzoic acid
PubChem CID 2735444
IUPAC Name 4-(benzimidazol-1-ylmethyl)benzoic acid
SMILES C1=CC=C2C(=C1)N=CN2CC3=CC=C(C=C3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 252.27
Percent Purity 97%
Chemical Name or Material 4-(1H-Benzimidazol-1-ylmethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.