missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1H-Benzimidazol-1-ylmethyl)benzoic acid, 97%, Thermo Scientific™
£47.20 - £124.00
Chemical Identifiers
CAS | 139742-50-0 |
---|---|
Molecular Formula | C15H12N2O2 |
Molecular Weight (g/mol) | 252.273 |
MDL Number | MFCD02180372 |
InChI Key | KLIANJXCGMBICV-UHFFFAOYSA-N |
Synonym | 4-1h-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzo d imidazol-1-yl methyl benzoic acid, 4-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-yl methyl benzoic acid, maybridge1_005258, acmc-20e4o9, 4-benzimidazolylmethyl benzoic acid, 4-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzimidazole-1-ylmethyl benzoic acid |
PubChem CID | 2735444 |
IUPAC Name | 4-(benzimidazol-1-ylmethyl)benzoic acid |
SMILES | C1=CC=C2C(=C1)N=CN2CC3=CC=C(C=C3)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10334342
|
Thermo Scientific Maybridge
KM10745DA |
1 g |
£47.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10049381
|
Thermo Scientific Maybridge
KM10745EA |
10 g |
£124.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
139742-50-0 | |
252.273 | |
KLIANJXCGMBICV-UHFFFAOYSA-N | |
2735444 | |
C1=CC=C2C(=C1)N=CN2CC3=CC=C(C=C3)C(=O)O |
C15H12N2O2 | |
MFCD02180372 | |
4-1h-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzo d imidazol-1-yl methyl benzoic acid, 4-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-yl methyl benzoic acid, maybridge1_005258, acmc-20e4o9, 4-benzimidazolylmethyl benzoic acid, 4-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzimidazole-1-ylmethyl benzoic acid | |
4-(benzimidazol-1-ylmethyl)benzoic acid |
Specifications
139742-50-0 | |
97% | |
MFCD02180372 | |
KLIANJXCGMBICV-UHFFFAOYSA-N | |
4-(benzimidazol-1-ylmethyl)benzoic acid | |
2735444 | |
97% |
97.0 | |
C15H12N2O2 | |
4-1h-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzo d imidazol-1-yl methyl benzoic acid, 4-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-ylmethyl benzoic acid, 4-1h-1,3-benzodiazol-1-yl methyl benzoic acid, maybridge1_005258, acmc-20e4o9, 4-benzimidazolylmethyl benzoic acid, 4-benzimidazol-1-ylmethyl benzoic acid, 4-1h-benzimidazole-1-ylmethyl benzoic acid | |
C1=CC=C2C(=C1)N=CN2CC3=CC=C(C=C3)C(=O)O | |
252.273 | |
252.27 | |
4-(1H-Benzimidazol-1-ylmethyl)benzoic acid |