missing translation for 'onlineSavingsMsg'
Learn More

[4-(1H-1,2,4-Triazol-1-ylmethyl)phenyl]methanol, ≥97%, Thermo Scientific™

Product Code. 10670885
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10670885 1 g Amber Glass Bottle 1g
10371323 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10670885 Supplier Thermo Scientific Maybridge Supplier No. CC45709DA
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 160388-56-7
Molecular Formula C10H11N3O
Molecular Weight (g/mol) 189.218
MDL Number MFCD07772869
InChI Key XEOWXPICRQBFIT-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl phenyl methanol, 4-1,2,4-triazol-1-ylmethyl phenyl methanol, 4-1h-1,2,4-triazol-1-ylmethyl benzyl alcohol, 4-1,2,4-triazolylmethyl phenyl methan-1-ol, 4-1h-1,2,4-triazol-1-yl methyl phenyl methanol
PubChem CID 7162035
IUPAC Name [4-(1,2,4-triazol-1-ylmethyl)phenyl]methanol
SMILES C1=CC(=CC=C1CN2C=NC=N2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.22
Percent Purity 97%
Chemical Name or Material [4-(1H-1, 2, 4-triazol-1-ylmethyl)phenyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.