missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10219934
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10219934 5 g 5g
10177552 1 g 1g
10477934 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10219934 Supplier Thermo Scientific Maybridge Supplier No. CC45716DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 112809-25-3
Molecular Formula C10H8N4
Molecular Weight (g/mol) 184.202
MDL Number MFCD07368016
InChI Key HQLYWHSJALKYOV-UHFFFAOYSA-N
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzonitrile, 4-1h-1,2,4-triazol-1-yl methyl benzonitrile, 4-1h-1,2,4-triazolylmethyl benzonitrile, 4-1,2,4-triazol-1-ylmethyl benzonitrile, benzonitrile, 4-1h-1,2,4-triazol-1-ylmethyl, 4-1,2,4-triazol-1-yl methyl benzonitrile, 4-1-1,2,4-triazolyl methyl benzontrile, 4-1,2,4-triazolylmethyl benzenecarbonitrile, letrozole impurity c
PubChem CID 7537616
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzonitrile
SMILES C1=CC(=CC=C1CN2C=NC=N2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-triazol-1-ylmethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.