missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-triazol-1-ylmethyl)benzoic acid, ≥97%, Thermo Scientific™

Product Code. 10442294
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10442294 10 g 10g
10340943 250 mg 250mg
10167402 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10442294 Supplier Thermo Scientific Maybridge Supplier No. CC45701EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 160388-54-5
Molecular Formula C10H9N3O2
Molecular Weight (g/mol) 203.201
MDL Number MFCD07187900
InChI Key DJXUEFGFCMIONN-UHFFFAOYSA-N
PubChem CID 3159714
IUPAC Name 4-(1,2,4-triazol-1-ylmethyl)benzoic acid
SMILES C1=CC(=CC=C1CN2C=NC=N2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Synonym 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1h-1,2,4-triazol-1-yl methyl benzoic acid, 4-1,2,4-triazol-1-ylmethyl benzoic acid, 4-1,2,4 triazol-1-ylmethyl-benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl benzoic acid hydrochloride, 4-1,2,4-triazolylmethyl benzoic acid, 4-1h-1,2,4-triazole-1-ylmethyl benzoic acid, benzoic acid,4-1h-1,2,4-triazol-1-ylmethyl, benzoic acid, 4-1h-1,2,4-triazol-1-ylmethyl
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 4-(1H-1, 2, 4-triazol-1-ylmethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.