missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-yl)benzoic acid, 95%, Thermo Scientific™

Product Code. 10163384
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10163384 10 g 10g
10487802 250 mg 250mg
10722483 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10163384 Supplier Thermo Scientific Maybridge Supplier No. CC18601EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 162848-16-0
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.174
MDL Number MFCD02682060
InChI Key FOMQQGKCPYKKHQ-UHFFFAOYSA-N
PubChem CID 736531
IUPAC Name 4-(1,2,4-triazol-1-yl)benzoic acid
SMILES C1=CC(=CC=C1C(=O)O)N2C=NC=N2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Synonym 4-1h-1,2,4-triazol-1-yl benzoic acid, 4-1,2,4 triazol-1-yl-benzoic acid, 4-1,2,4-triazol-1-yl benzoic acid, benzoic acid, 4-1h-1,2,4-triazol-1-yl, benzoic acid,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzoic acid, pubchem19582, rarechem al be 1325, buttpark 9550-80, acmc-1c5ri
Formula Weight 189.17
Percent Purity 95%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.