CAS RN 162848-16-0
IUPAC Name:
4-(1H-1,2,4-triazol-1-yl)benzoate
Synonyms:
4-(1,2,4-triazol-1-yl)benzoate
Molecular Weight (g/mol):
188.17
Molecular Formula:
C9H6N3O2
InChi Key:
FOMQQGKCPYKKHQ-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1=CC=C(C=C1)N1C=NC=N1
1
–
1
of
1
results
1
4-(1H-1,2,4-Triazol-1-yl)benzoic acid, 95%, Thermo Scientific™
CAS: 162848-16-0 Molecular Formula: C9H7N3O2 Molecular Weight (g/mol): 189.174 MDL Number: MFCD02682060 InChI Key: FOMQQGKCPYKKHQ-UHFFFAOYSA-N PubChem CID: 736531 IUPAC Name: 4-(1,2,4-triazol-1-yl)benzoic acid SMILES: C1=CC(=CC=C1C(=O)O)N2C=NC=N2