missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-1,2,4-Triazol-1-yl)benzaldehyde, 95+%, Thermo Scientific™

Product Code. 10081474 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081474 5 g Amber Glass Bottle 5g
10099221 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081474 Supplier Thermo Scientific Maybridge Supplier No. CC18604DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 27996-86-7
Molecular Formula C9H7N3O
Molecular Weight (g/mol) 173.175
InChI Key TVEJNWMWDIXPAX-UHFFFAOYSA-N
PubChem CID 2776488
IUPAC Name 4-(1,2,4-triazol-1-yl)benzaldehyde
SMILES C1=CC(=CC=C1C=O)N2C=NC=N2

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 4-1h-1,2,4-triazol-1-yl benzaldehyde, 4-1,2,4 triazol-1-yl-benzaldehyde, 4-1,2,4-triazol-1-yl benzaldehyde, 4-1h-1,2,4-triazol-1-yl benzenecarbaldehyde, benzaldehyde, 4-1h-1,2,4-triazol-1-yl, 1-4-formylphenyl-1,2,4-triazole, benzaldehyde,4-1h-1,2,4-triazol-1-yl, 4-1,2,4-triazolyl benzaldehyde, 4-1,2,4-triazol-1-yl-benzaldehyde
Formula Weight 173.17
Percent Purity 95%
Chemical Name or Material 4-(1H-1, 2, 4-Triazol-1-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.