missing translation for 'onlineSavingsMsg'
Learn More

4-1BB/TNFRSF9/CD137 Mouse anti-Human, Clone: 4B4-1-1, Novus Biologicals™

Product Code. 18384583
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384583 0.025 mg 25µg
18369633 0.2 mg 0.2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18384583 Supplier Novus Biologicals Supplier No. NBP3120280.025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Applications Flow Cytometry, ELISA, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 4B4-1-1
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA, Immunohistochemistry, Block/Neutralize
Formulation PBS
Gene Alias 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Host Species Mouse
Immunogen The original antibody was generated by immunizing BALB/c mice with a fusion protein of human 4-1BB/TNFRSF9/CD137 antigen and glutathione s transferase (GST).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3604
Target Species Human
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.