missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-1BB/TNFRSF9/CD137 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £310.00
Specifications
| Antigen | 4-1BB/TNFRSF9/CD137 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161893
|
Novus Biologicals
NBP2-31365SS |
0.025 mg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159084
|
Novus Biologicals
NBP2-31365 |
0.1 mg |
£310.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-1BB/TNFRSF9/CD137 Polyclonal specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot.Specifications
| 4-1BB/TNFRSF9/CD137 | |
| Western Blot | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Q07011 | |
| 3604 | |
| CD137 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Stem Cells | |
| PBS with 0.05% Sodium Azide | |
| 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9 | |
| TNFRSF9 | |
| IgG | |
| Protein A purified | |
| Human. The immunogen shows 63% similarity with CD137 of Mouse. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title