missing translation for 'onlineSavingsMsg'
Learn More

4-1BB/TNFRSF9/CD137 Antibody (4B4-1-1), Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18353693 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18353693 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18353693 Supplier Novus Biologicals Supplier No. NBP312029AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

4-1BB/TNFRSF9/CD137 Monoclonal antibody specifically detects 4-1BB/TNFRSF9/CD137 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BB/TNFRSF9/CD137
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 4B4-1-1
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 4-1BB, 4-1BB ligand receptor, CD137 antigen, CD137MGC2172, CDw137, FLJ43501, ILAhomolog of mouse 4-1BB, induced by lymphocyte activation (ILA), interleukin-activated receptor, homolog of mouse Ly63, receptor protein 4-1BB, T cell antigen ILA, T-cell antigen 4-1BB homolog, T-cell antigen ILA, tumor necrosis factor receptor superfamily member 9, tumor necrosis factor receptor superfamily, member 9
Host Species Rabbit
Immunogen The original antibody was generated by immunizing BALB/c mice with a fusion protein of human 4-1BB/TNFRSF9/CD137 antigen and glutathione s transferase (GST).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immune System Diseases, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3604
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.