missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-1BB Ligand/TNFSF9, Rabbit anti-Mouse, Clone: TKS-1, Novus Biologicals™
Rabbit Monoclonal Antibody
£207.00 - £463.00
Specifications
| Antigen | 4-1BB Ligand/TNFSF9 |
|---|---|
| Clone | TKS-1 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry, Block/Neutralize |
| Applications | Flow Cytometry, Blocking Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18384221
|
Novus Biologicals
NBP2-81115-0.025MG |
0.025 mg |
£207.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329190
|
Novus Biologicals
NBP2-81115-0.2MG |
0.2 mg |
£463.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, ELISA, Block/Neutralize.Specifications
| 4-1BB Ligand/TNFSF9 | |
| 1 mg/mL | |
| Flow Cytometry, Blocking Assay | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins | |
| PBS | |
| 8744 | |
| Murine B-cell lymphoma 2PK-3 cells injected into SD rats. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| TKS-1 | |
| Flow Cytometry, Block/Neutralize | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 | |
| TNFSF9 | |
| IgG κ | |
| Protein A purified | |
| This antibody binds to murine 4-1BB Ligand/TNFSF9 and competes with binding of 4-1BB. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title