missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-1BB Ligand/TNFSF9 Antibody (TKS-1), Alexa Fluor™ 405, Novus Biologicals™
Shop All Bio Techne ProductsDescription
4-1BB Ligand/TNFSF9 Monoclonal specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, ELISA.
Specifications
Specifications
| Antigen | 4-1BB Ligand/TNFSF9 |
| Applications | Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Clone | TKS-1 |
| Conjugate | Alexa Fluor 405 |
| Dilution | Flow Cytometry, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9 |
| Gene Symbols | TNFSF9 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?