missing translation for 'onlineSavingsMsg'
Learn More

4-1BB Ligand/TNFSF9 Antibody (AT113-2), DyLight 350, Novus Biologicals™

Product Code. 18377422 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377422 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377422 Supplier Novus Biologicals Supplier No. NBP311987UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

4-1BB Ligand/TNFSF9 Monoclonal antibody specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-1BB Ligand/TNFSF9
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone AT113-2
Conjugate DyLight 350
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias 4-1BB ligand, 4-1BBL, 4-1BB-L, CD137L, homolog of mouse 4-1BB-L, receptor 4-1BB ligand, tumor necrosis factor (ligand) superfamily, member 9, tumor necrosis factor ligand superfamily member 9
Host Species Rat
Immunogen This antibody was raised by immunising LOU rats with soluble recombinant 4-1BB Ligand/TNFSF9:Fc fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 8744
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.