Learn More
Invitrogen™ 4-1BB (CD137/TNFRSF9) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMTNFRSF9X5
Additional Details : Weight : 0.33800kg
Description
The Mouse 4-1BB (CD137/TNFRSF9) ELISA quantitates Ms 4-1BB in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms 4-1BB. Principle of the method The Mouse 4-1BB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CD137 is a member of the TNF-receptor superfamily and contributes to the clonal expansion, survival, and development of T cells. CD137 exists as both a monomer and a dimer on the surface of activated T cells. CD137 can also induce proliferation in peripheral monocytes, enhance T cell apoptosis induced by TCR/CD3 triggered activation, and regulate CD28 co-stimulation to promote Th1 cell responses. The expression of CD137 is induced by lymphocyte activation. CD137 is also an inducible receptor-like protein expressed on the cell surface of activated splenic T cells and thymocytes. CD137 exists as both a monomer and a dimer on the surface of activated T cells.Specifications
P20334 | |
6 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
21942 | |
<12% | |
Wash Buffer, Pre-coated 96 well plates, Assay Diluent Concentrate, Adhesive Plate Covers, Standard, Chromogen, Stop Solution, SAV-HRP, Biotinylated Detection Antibody | |
5 x 96 Tests | |
Immunology | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
6.14 pg/mL to 1500 pg/mL | |
6 pg/mL to 1500 pg/mL | |
Biotin | |
Cytokines & Receptors | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN-gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-4, IL-13, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
4-1 BB,A930040I11Rik,AA408498,AI325004,CDw137,Cd137,ILA,Ly63 | |
<10% | |
HRP | |
RUO | |
Plasma, 33 μ;L; Serum, 33 μ;L; Supernatant, 100 μ;L | |
RP23-272N19.6, 4-1 BB, A930040I11Rik, AA408498, AI325004, CDw137, Cd137, ILA, Ly63 | |
1 hr. 20 min. |