missing translation for 'onlineSavingsMsg'
Learn More

4,17-Dioxabicyclo[12.3.2]nonadecane-18-O-tert-butyldimethylsilyl Spiramycin I-d3, TRC

Product Code. 30378390
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30378390 10 mg 10mg
30375080 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30378390 Supplier Toronto Research Chemicals Supplier No. TRCD48525210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,17-Dioxabicyclo[12.3.2]nonadecane-18-O-tert-butyldimethylsilyl Spiramycin I-d3
Molecular Formula C49D3H85N2O14Si
Quantity 10 mg
Synonym [9(2R,5S,6R)]-18-Deoxo-3-deoxy-9-O-[5-(dimethylamino)tetrahydro-6-methyl-2H-pyran-2-yl]-18-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-3,18-epoxy-leucomycin V
InChI Formula InChI=1S/C49H88N2O14Si/c1-15-16-24-66(13,14)65-40-26-34-25-29(2)36(61-39-23-22-35(50(8)9)31(4)58-39)21-19-17-18-20-30(3)57-38(52)27-37(62-40)46(56-12)45(34)64-48-43(53)42(51(10)11)44(32(5)60-48)63-41-28-49(7,55)47(54)33(6)59-41/h17-19,21,29-37,39-48,53-55H,15-16,20,22-28H2,1-14H3/b18-17+,21-19+/t29-,30-,31?,32?,33-,34-,35+,36+,37-,39+,40?,41+,42?,43?,44-,45+,46+,47+,48+,49?/m1/s1/i8D3
SMILES [2H]C([2H])([2H])N(C)[C@H]1CC[C@H](O[C@H]2\C=C\C=C\C[C@@H](C)OC(=O)C[C@H]3OC(C[C@@H](C[C@H]2C)[C@H](O[C@@H]4OC(C)[C@@H](O[C@H]5CC(C)(O)[C@@H](O)[C@@H](C)O5)C(C4O)N(C)C)[C@H]3OC)O[Si](C)(C)CCCC)OC1C
IUPAC Name (1R,3R,4R,5E,7E,10R,14R,18S,19S)-16-[butyl(dimethyl)silyl]oxy-18-[(2S,5S)-5-[(2S,5S,6R)-4,5-dihydroxy-4,6-dimethyl-tetrahydropyran-2-yl]oxy-4-(dimethylamino)-3-hydroxy-6-methyl-tetrahydropyran-2-yl]oxy-19-methoxy-3,10-dimethyl-4-[(2R,5S)-6-methyl-5-[methyl(trideuteriomethyl)amino]tetrahydropyran-2-yl]oxy-11,15-dioxabicyclo[12.3.2]nonadeca-5,7-dien-12-one
Molecular Weight (g/mol) 960.33
Formula Weight 959.6193
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.