missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzoic acid, Thermo Scientific™

Product Code. 10155442
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 22090-27-3
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD01631241
InChI Key KPCBFFYRSJPCJH-UHFFFAOYSA-N
PubChem CID 2795515
SMILES OC(=O)C1=CC=C(C=C1)N1CCCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-pyrrolidin-1-yl benzoic acid, 4-1-pyrrolidinyl benzoic acid, 4-pyrrolidin-1-yl-benzoic acid, 4-1-pyrrolidinyl benzenecarboxylic acid, benzoic acid,4-1-pyrrolidinyl, benzoic acid, 4-1-pyrrolidinyl, 4-pyrrolidinylbenzoic acid, pubchem19562, 1-4-carboxyphenyl pyrrolidine
IUPAC Name 4-(pyrrolidin-1-yl)benzoic acid
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'