Learn More
4-(1-Pyrrolidinyl)benzoic acid, Maybridge™
Brand: Maybridge CC30101DA
Code : 31
Additional Details : CAS Number : 22090-27-3 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
22090-27-3 | |
191.23 | |
4-pyrrolidin-1-yl benzoic acid, 4-1-pyrrolidinyl benzoic acid, 4-pyrrolidin-1-yl-benzoic acid, 4-1-pyrrolidinyl benzenecarboxylic acid, benzoic acid,4-1-pyrrolidinyl, benzoic acid, 4-1-pyrrolidinyl, 4-pyrrolidinylbenzoic acid, pubchem19562, 1-4-carboxyphenyl pyrrolidine | |
4-pyrrolidin-1-ylbenzoic acid |
C11H13NO2 | |
KPCBFFYRSJPCJH-UHFFFAOYSA-N | |
2795515 | |
C1CCN(C1)C2=CC=C(C=C2)C(=O)O |
Specifications
4-(1-Pyrrolidinyl)benzoic acid | |
1g | |
97% | |
4-pyrrolidin-1-yl benzoic acid, 4-1-pyrrolidinyl benzoic acid, 4-pyrrolidin-1-yl-benzoic acid, 4-1-pyrrolidinyl benzenecarboxylic acid, benzoic acid,4-1-pyrrolidinyl, benzoic acid, 4-1-pyrrolidinyl, 4-pyrrolidinylbenzoic acid, pubchem19562, 1-4-carboxyphenyl pyrrolidine | |
C1CCN(C1)C2=CC=C(C=C2)C(=O)O | |
191.23 | |
191.23 |
Amber Glass Bottle | |
22090-27-3 | |
C11H13NO2 | |
KPCBFFYRSJPCJH-UHFFFAOYSA-N | |
4-pyrrolidin-1-ylbenzoic acid | |
2795515 | |
97% |