missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester, 97%

Product Code. 15425346
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425346 1 g 1g
15435346 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425346 Supplier Thermo Scientific Alfa Aesar Supplier No. H59911.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 852227-90-8 | C16H24BNO2 | 273.18 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 852227-90-8
Molecular Formula C16H24BNO2
Molecular Weight (g/mol) 273.18
MDL Number MFCD08060504
InChI Key DWJNNJSONWFVBT-UHFFFAOYSA-N
Synonym 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid pinacol ester, 4-1-pyrrolidinyl benzeneboronic acid pinacol ester, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrrolidine, 4-pyrrolidinophenylboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenylboronic acid pinacol ester, 4-pyrrolidin-1-yl benzeneboronic acid, pinacol ester, 4-pyrrolidin-1-yl phenyl boronic acid pinacol ester, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl phenyl pyrrolidine
PubChem CID 18525715
IUPAC Name 1-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)N1CCCC1

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 273.18
Percent Purity 97%
Chemical Name or Material 4-(1-Pyrrolidinyl)benzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.