missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Pyrrolidinyl)-1-butylamine, 98%

Product Code. 11419490
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419490 1 g 1g
11429490 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419490 Supplier Thermo Scientific Alfa Aesar Supplier No. H50323.03
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 24715-90-0 | C8H18N2 | 142.25 g/mol

4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Pyrrolidinyl)-1-butylamine is used directly as drugs, food, cosmetics. Used as a building block in organic synthesis, pyrrolidine activates ketones and aldehydes toward nucleophilic addition by formation of enamines

Solubility
Soluble in water

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24715-90-0
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.25
MDL Number MFCD00192591
InChI Key LSDYCEIPEBJKPT-UHFFFAOYSA-N
Synonym 4-pyrrolidin-1-yl butan-1-amine, 4-1-pyrrolidinyl-1-butanamine, 1-pyrrolidinebutanamine, 4-1-pyrrolidino butylamine, 4-pyrrolidinobutylamine, 4-1-pyrrolidinyl-1-butylamine, 1-4-aminobutyl pyrrolidine, 4-pyrrolidin-1-yl-butylamine, 4-pyrrolidinylbutylamine, 1-pyrrolidinebutylamine
PubChem CID 2762762
IUPAC Name 4-pyrrolidin-1-ylbutan-1-amine
SMILES NCCCCN1CCCC1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in water
Formula Weight 142.25
Percent Purity 98%
Chemical Name or Material 4-(1-Pyrrolidinyl)-1-butylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.