missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methylethyl)benzoic Acid, TRC

Product Code. 30466722
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466722 10 g 10g
30476529 25 g 25g
30466130 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30466722 Supplier Toronto Research Chemicals Supplier No. TRCM30485510G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-66-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
Synonym p-Isopropylbenzoic Acid, 4-(1-Methylethyl)benzoic Acid, 4-Isopropylbenzoic Acid, Cumic Acid, Cuminic Acid, NSC 1907, NSC 20083, p-Isopropylbenzoic Acid
IUPAC Name 4-propan-2-ylbenzoic acid
SMILES CC(C)c1ccc(cc1)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H12O2/c1-7(2)8-3-5-9(6-4-8)10(11)12/h3-7H,1-2H3,(H,11,12)
Formula Weight 164.0837
Chemical Name or Material 4-(1-Methylethyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.