missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10602515
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10602515 1 g Amber Glass Bottle 1g
11320587 250 mg Amber Glass Bottle 250mg
2 options
This item is not returnable. View return policy

Product Code. 10602515

Brand: Thermo Scientific Maybridge CC23803DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 916766-81-9
Molecular Formula C10H9ClN2O2S
Molecular Weight (g/mol) 256.704
MDL Number MFCD09817463
InChI Key PXFXKTMOZHYJJU-UHFFFAOYSA-N
Synonym 4-1-methyl-1h-pyrazol-3-yl benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl-benzenesulfonyl chloride, 4-1-methylpyrazol-3-yl benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl benzenesulphonyl chloride, chloro 4-1-methylpyrazol-3-yl phenyl sulfone, 3-4-chlorosulphonyl phenyl-1-methyl-1h-pyrazole, 4-1-methyl-1h-pyrazol-3-yl phenylsulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methyl-1h-pyrazol-3-yl
PubChem CID 24229467
IUPAC Name 4-(1-methylpyrazol-3-yl)benzenesulfonyl chloride
SMILES CN1C=CC(=N1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 256.71
Percent Purity 95%
Chemical Name or Material 4-(1-Methyl-1H-pyrazol-3-yl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.