missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methyl-1H-pyrazol-3-yl)aniline, 97%, Thermo Scientific™

Product Code. 10413204
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10413204 250 mg Amber Glass Bottle 250mg
10286483 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10413204 Supplier Thermo Scientific Maybridge Supplier No. CC23814CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 916766-82-0
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.219
MDL Number MFCD03407387
InChI Key WEFJHPBVOKVCLZ-UHFFFAOYSA-N
PubChem CID 18526375
IUPAC Name 4-(1-methylpyrazol-3-yl)aniline
SMILES CN1C=CC(=N1)C2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-1-methyl-1h-pyrazol-3-yl aniline, 4-1-methyl-1h-pyrazol-3-yl-phenylamine, 4-1-methylpyrazol-3-yl aniline, 1-methyl-3-4-aminophenyl-1h-pyrazole, benzenamine, 4-1-methyl-1h-pyrazol-3-yl, 4-1-methylpyrazol-3-yl phenylamine, 4-1-methyl-1h-pyrazol-3-yl-aniline, 4-1-methyl-1h-pyrazol-3-yl phenylamine, benzenamine,4-1-methyl-1h-pyrazol-3-yl
Formula Weight 173.22
Percent Purity 97%
Chemical Name or Material 4-(1-Methyl-1H-pyrazol-3-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.