missing translation for 'onlineSavingsMsg'
Learn More

4-{1-Hydroxy-2-[3-(4-hydroxyphenyl)-5-methylcyclohexylamino]ethyl}phenol Hydrochloride (Mixture of Diastereomers), TRC

Product Code. 30407987
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407987 1 mg 1mg
30393716 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407987 Supplier Toronto Research Chemicals Supplier No. TRCH8253901MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[1-Hydroxy-2-[3-(4-hydroxyphenyl)-5-methylcyclohexylamino]ethyl]phenol hydrochloride (mixture of diastereomers)
Analyte or Component Names 4-[1-Hydroxy-2-[3-(4-hydroxyphenyl)-5-methylcyclohexylamino]ethyl]phenol hydrochloride (mixture of diastereomers)
Shipping Condition Room Temperature
Molecular Formula C21 H27 N O3 . H Cl
InChI Formula InChI=1S/C21H27NO3.ClH/c1-14-10-17(15-2-6-19(23)7-3-15)12-18(11-14)22-13-21(25)16-4-8-20(24)9-5-16;/h2-9,14,17-18,21-25H,10-13H2,1H3;1H
Solution Type Neat
SMILES Cl.CC1CC(CC(C1)c2ccc(O)cc2)NCC(O)c3ccc(O)cc3
IUPAC Name 4-[3-[[2-hydroxy-2-(4-hydroxyphenyl)ethyl]amino]-5-methyl-cyclohexyl]phenol;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 377.9
Quantity 1 mg
Formula Weight 377.1758
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.