missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Cyclopenten-1-yl)morpholine, 96%, Thermo Scientific™

Product Code. p-7043329
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11379963

Brand: Thermo Scientific Alfa Aesar L00262.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-52-7
Molecular Formula C9H15NO
Molecular Weight (g/mol) 153.225
MDL Number MFCD00006162
InChI Key VAPOFMGACKUWCI-UHFFFAOYSA-N
Synonym 1-morpholinocyclopentene, n-1-cyclopenten-1-yl morpholine, 1-morpholino-1-cyclopentene, morpholine, 4-1-cyclopenten-1-yl, 4-1-cyclopenten-1-yl morpholine, 4-1-cyclopentenyl morpholine, 1-n-morpholino cyclopentene, n-1-cyclopenten-1-yl-morpholine, 4-cyclopent-1-en-1-yl morpholine
PubChem CID 70294
IUPAC Name 4-(cyclopenten-1-yl)morpholine
SMILES C1CC=C(C1)N2CCOCC2

Specifications

Density 0.957
Boiling Point 105°C to 106°C (12 mmHg)
Flash Point 64°C (147°F)
Refractive Index 1.5118
Quantity 25 g
Beilstein 117154
Formula Weight 153.23
Percent Purity 96%
Chemical Name or Material 4-(1-Cyclopenten-1-yl)morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.