missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Bromoethyl)benzoic acid, 98%

Product Code. 11357846
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357846 1 g 1g
11367846 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357846 Supplier Thermo Scientific Alfa Aesar Supplier No. L13025.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 113023-73-7 | C9H9BrO2 | 229.073 g/mol

4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113023-73-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00020378
InChI Key VICCYULHZWEWMB-UHFFFAOYSA-N
Synonym 4-1-bromoethyl benzoic acid, benzoic acid, 4-1-bromoethyl, acmc-1c3tf, 4-1-bromoethyl-benzoic acid, 4-1-bromo-ethyl-benzoic acid, benzoic acid,4-1-bromoethyl
PubChem CID 5146225
IUPAC Name 4-(1-bromoethyl)benzoic acid
SMILES CC(C1=CC=C(C=C1)C(=O)O)Br

Specifications

Melting Point 155°C to 156°C
Quantity 1 g
UN Number UN3261
Beilstein 2084039
Solubility Information Insoluble in water.
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 4-(1-Bromoethyl)benzoic acid

RUO – Research Use Only

Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.