missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Bromoethyl)-6-chloro-5-fluoropyrimidine, Mikromol™

Product Code. 30051634
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051634 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051634 Supplier Mikromol Supplier No. MM1094.130100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-(1-Bromoethyl)-6-chloro-5-fluoropyrimidine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(1-Bromoethyl)-6-chloro-5-fluoropyrimidine
CAS 188416-28-6
Analyte or Component Names 4-(1-Bromoethyl)-6-chloro-5-fluoropyrimidine
Shipping Condition Room Temperature
Molecular Formula C6 H5 Br Cl F N2
Certifications/Compliance ISO 9001
Synonym Pyrimidine, 4-(1-bromoethyl)-6-chloro-5-fluoro-, 4-(1-Bromoethyl)-6-chloro-5-fluoropyrimidine
InChI Formula InChI=1S/C6H5BrClFN2/c1-3(7)5-4(9)6(8)11-2-10-5/h2-3H,1H3
SMILES CC(Br)c1ncnc(Cl)c1F
IUPAC Name 4-(1-bromoethyl)-6-chloro-5-fluoro-pyrimidine
Molecular Weight (g/mol) 239.473
Quantity 100 mg
Formula Weight 237.9309 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.