missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(1,4-Cyclohexadien-1-yl)-1-butanol, 97%, stab. with 0.1% BHT

Product Code. 15479065
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479065 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15479065 Supplier Alfa Aesar™ Supplier No. H36058.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 956150-13-3
Molecular Formula C10H16O
Molecular Weight (g/mol) 152.237
MDL Number MFCD22682825
InChI Key QUBPGTNRIJGDFP-UHFFFAOYSA-N
Synonym 4-cyclohexa-1,4-dien-1-yl butan-1-ol, 4-1,4-cyclohexadien-1-yl-1-butanol
PubChem CID 57103928
IUPAC Name 4-cyclohexa-1,4-dien-1-ylbutan-1-ol
SMILES C1C=CCC(=C1)CCCCO

Specifications

Density 0.957
Boiling Point 247°C to 248°C
Flash Point 103°C (217°F)
Refractive Index 1.503
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 152.23
Percent Purity 97%
Chemical Name or Material 4-(1,4-Cyclohexadien-1-yl)-1-butanol, stabilized with 0.1% BHT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.