missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Thiazol-2-yl)benzaldehyde, 95%, Thermo Scientific™

Product Code. 10439953
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10439953 1 g 1g
10576314 250 mg 250mg
10193214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10439953 Supplier Thermo Scientific Maybridge Supplier No. CC40204DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 198904-53-9
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.23
MDL Number MFCD06797780
InChI Key LQLBILPEELCFQI-UHFFFAOYSA-N
PubChem CID 10535655
IUPAC Name 4-(1,3-thiazol-2-yl)benzaldehyde
SMILES O=CC1=CC=C(C=C1)C1=NC=CS1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 4-thiazol-2-yl-benzaldehyde, 4-1,3-thiazol-2-yl benzaldehyde, 4-thiazol-2-yl benzaldehyde, benzaldehyde,4-2-thiazolyl, benzaldehyde, 4-2-thiazolyl, 4-2-thiazolyl benzaldehyde, 4-thiazol-2-ylbenzaldehyde, 4-2-thiazolyl-benzaldehyde
Formula Weight 189.24
Percent Purity 95%
Chemical Name or Material 4-(1, 3-Thiazol-2-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.