missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Oxazol-5-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10702663
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10702663 250 mg Amber Glass Bottle 250mg
10633783 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10702663 Supplier Thermo Scientific Maybridge Supplier No. CC14501CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 250161-45-6
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD02682033
InChI Key MCPBXFWYUHYSIZ-UHFFFAOYSA-N
Synonym 4-1,3-oxazol-5-yl benzoic acid, 4-oxazol-5-yl benzoic acid, 4-5-oxazolyl benzoic acid, benzoic acid, 4-5-oxazolyl, 4-oxazol-5-ylbenzoic acid, 4-5-oxazolyl benzoicacid, 4-oxazol-5-yl-benzoic acid, benzoic acid,4-5-oxazolyl, 4-l,3-oxazol-5-yl benzoic acid
PubChem CID 2776414
IUPAC Name 4-(1,3-oxazol-5-yl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=CN=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.