missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Oxazol-5-yl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10742093
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10742093 1 g Amber Glass Bottle 1g
10292342 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10742093 Supplier Thermo Scientific Maybridge Supplier No. CC14503DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-66-4
Molecular Formula C9H6ClNO3S
Molecular Weight (g/mol) 243.66
MDL Number MFCD02681962
InChI Key QCGUTLAPXWBIMM-UHFFFAOYSA-N
Synonym 4-1,3-oxazol-5-yl benzenesulfonyl chloride, 4-oxazol-5-yl benzene-1-sulfonyl chloride, 4-oxazol-5-yl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-5-oxazolyl, 4-1,3-oxazol-5-yl benzenesulfonylchloride, 4-1,3-oxazol-5-yl benzene-1-sulfonyl chloride, 4-1,3-oxazol-5-yl benzenesulphonyl chloride, 4-oxazol-5-ylbenzenesulfonyl-chloride, 4-oxazol-5-ylbenzene-sulfonyl-chloride, chloro 4-1,3-oxazol-5-yl phenyl sulfone
PubChem CID 2776415
IUPAC Name 4-(1,3-oxazol-5-yl)benzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1=CN=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 243.67
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.