missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Oxazol-5-yl)aniline, 97%, Thermo Scientific™

Product Code. p-2700393
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10763433 250 mg Amber Glass Bottle 250mg
10202982 1 g Amber Glass Bottle 1g
10240515 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10763433 Supplier Thermo Scientific Maybridge Supplier No. DFP00120CB
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1008-95-3
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.176
MDL Number MFCD00052192
InChI Key SLJBMRSOKUTXDF-UHFFFAOYSA-N
PubChem CID 517747
IUPAC Name 4-(1,3-oxazol-5-yl)aniline
SMILES C1=CC(=CC=C1C2=CN=CO2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-1,3-oxazol-5-yl aniline, 4-5-oxazolyl aniline, 4-oxazol-5-yl aniline, benzenamine, 4-5-oxazolyl, 5-4-aminophenyl oxazole, benzenamine,4-5-oxazolyl, 4-oxazol-5-yl-aniline, maybridge3_001299, 4-oxazol-5-yl-phenylamine
Formula Weight 160.18
Percent Purity 97%
Chemical Name or Material 4-(1, 3-Oxazol-5-yl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.