missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3-Dioxolan-2-yl)piperidine, ≥90%, Thermo Scientific™

Product Code. 10485512
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10485512 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10485512 Supplier Thermo Scientific Maybridge Supplier No. MO00374CB
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 423768-60-9
CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C13H23NO6
MDL Number MFCD03659711
Quantity 250 mg
Synonym 4-5,5-dimethyl-1,3-dioxan-2-yl piperidine oxalate, 4-5,5-dimethyl-1,3-dioxan-2-yl piperidine; oxalic acid
InChI Key VIVKYBWHPORZOA-UHFFFAOYSA-N
SMILES CC1(COC(OC1)C2CCNCC2)C.C(=O)(C(=O)O)O
IUPAC Name 4-(5,5-dimethyl-1,3-dioxan-2-yl)piperidine;oxalic acid
Molecular Weight (g/mol) 289.328
PubChem CID 2779761
Formula Weight 289.33
Percent Purity 90%
Chemical Name or Material 4-(5, 5-Dimethyl-1, 3-dioxan-2-yl)piperidine oxalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.