missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(1,3-Dimethylbutylamino)diphenylamine, TRC
Specifications
Specifications
| Chemical Name or Material | N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine |
| CAS | 793-24-8 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H24 N2 |
| Quantity | 10 g |
| Synonym | 1,4-Benzenediamine, N-(1,3-dimethylbutyl)-N'-phenyl- (9CI), p-Phenylenediamine, N-(1,3-dimethylbutyl)-N'-phenyl- (7CI,8CI), N1-(1,3-Dimethylbutyl)-N4-phenyl-1,4-benzenediamine, 1-N-(4-Methylpentan-2-yl)-4-N-phenylbenzene-1,4-diamine, 4-(1,3-Dimethylbutylamino)diphenylamine, 6PPD, Accinox ZC, Antage 6C, Antigene 6C, Antioxidant 4020, Antioxidant 6C, Antioxidant CD 13, Antioxidant PD 2, Antozite 67, Antozite 67F, CD 13, CI, DBDA, DMBPD, Diafen 13, Diafen FDMB, Dusantox 6PPD, Flexzone 7F, Flexzone 7L, Flexzone 7P, Forte 6C, Kumanox 13, Kumanox 13C, Luvomaxx 6PPD, N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-benzenediamine, N-(1,3-Dimethylbutyl)-N'-phenyl-1,4-phenylenediamine, N-(1,3-Dimethylbutyl)-N'-phenyl-p-phenylenediamine, N-(4-Methyl-2-pentyl)-N'-phenyl-p-phenylenediamine, N-Phenyl-N'-(1,3-dimethylbutyl)-1,4-phenylenediamine, N-Phenyl-N'-(1,3-dimethylbutyl)-p-phenylenediamine, Nocceler 6C, Nocrac 6C, Ozonone 6C, PD 2, Permanax 120, Permanax 6PPD, Pilflex 13, Santoflex 13, Santoflex 13F, Santoflex 6PPD, Sunsine 6PPD, UOP 562, UOP 588, Vulkanox 4020, Vulkanox 4020LG, Wingstay 300, 1,4-Benzenediamine, N1-(1,3-dimethylbutyl)-N4-phenyl- |
| InChI Formula | InChI=1S/C18H24N2/c1-14(2)13-15(3)19-17-9-11-18(12-10-17)20-16-7-5-4-6-8-16/h4-12,14-15,19-20H,13H2,1-3H3 |
| SMILES | CC(C)CC(C)Nc1ccc(Nc2ccccc2)cc1 |
| IUPAC Name | 1-N-(4-methylpentan-2-yl)-4-N-phenylbenzene-1,4-diamine |
| Molecular Weight (g/mol) | 268.4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?