missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,3-Thiadiazol-4-yl)benzylamine hydrochloride, ≥95%, Thermo Scientific™

Product Code. 10475692
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10475692 1 g Amber Glass Bottle 1g
10089101 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10475692 Supplier Thermo Scientific Maybridge Supplier No. CC16413DA
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-49-9
Molecular Formula C9H10ClN3S
Molecular Weight (g/mol) 227.71
MDL Number MFCD02682075
InChI Key WUUGGOQPXZDQNW-UHFFFAOYSA-N
Synonym 4-1,2,3-thiadiazol-4-yl benzylamine hydrochloride, 4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 1-4-1,2,3-thiadiazol-4-yl phenyl methanamine hydrochloride, 4-thiadiazol-4-yl phenyl methanamine hydrochloride, benzenemethanamine,4-1,2,3-thiadiazol-4-yl, 4-1,2,3-thiadiazol-4-yl phenyl methylamine, chloride, 4-4-aminomethyl phenyl-1,2,3-thiadiazole hydrochloride
PubChem CID 2737287
IUPAC Name [4-(thiadiazol-4-yl)phenyl]methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)C2=CSN=N2.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 227.72
Percent Purity 95%
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.