missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetic Acid, Mikromol™
Description
[4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetic Acid
Specifications
Specifications
| Chemical Name or Material | (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetic Acid |
| CAS | 854646-92-7 |
| Analyte or Component Names | (4-(1,1-Dimethylethyl)-2,6-dimethylphenyl)acetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H20 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Xylometazoline Hydrochloride Imp. F (EP), Xylometazoline Imp. F (EP), [4-(1,1-Dimethylethyl)-2,6-dimethylphenyl]acetic Acid, Xylometazoline Hydrochloride Impurity F, Xylometazoline Impurity F |
| InChI Formula | InChI=1S/C14H20O2/c1-9-6-11(14(3,4)5)7-10(2)12(9)8-13(15)16/h6-7H,8H2,1-5H3,(H,15,16) |
| SMILES | Cc1cc(cc(C)c1CC(=O)O)C(C)(C)C |
| IUPAC Name | 2-(4-tert-butyl-2,6-dimethylphenyl)acetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?