Learn More
WTW™ 4.01 pH Technical Buffer Solution
GREENER_CHOICE
Product Code. 10716645
Click to view available options
Type:
Technical Buffer Solution
Unit Size:
Each
Product Code. 10716645
Brand: WTW™ 108701
Precise and traceable to PTB/NIST with built-in dosing vessel standard buffer.
  • Life up to 3 years unopened
  • Thrifty use
  • Easiest handling by means of dosing device
  • Supplied in 1L bottles
TRUSTED_SUSTAINABILITY
Specifications
Description 4.01 pH
Type Technical Buffer Solution
Size 1L