missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid (Penillic Acid of Benzylpenicillin), Mikromol™
Description
(3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid (Penillic Acid of Benzylpenicillin)
Specifications
Specifications
| Chemical Name or Material | (3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid (Penillic Acid of Benzylpenicillin) |
| CAS | 13093-87-3 |
| Analyte or Component Names | (3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid (Penillic Acid of Benzylpenicillin) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H18 N2 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzylpenicillin Benzathine Imp. D (EP), Benzylpenicillin Potassium Imp. D (EP), Benzylpenicillin Procaine Imp. D (EP), (3S,7R,7aR)-5-Benzyl-2,2-dimethyl-2,3,7,7a-tetrahydroimidazo[5,1-b]thiazole-3,7-dicarboxylic Acid, Penillic Acid of Benzylpenicillin, Benzylpenicillin Benzathine Impurity D, Benzylpenicillin Potassium Impurity D, Benzylpenicillin Procaine Impurity D, Carbenicillin Sodium Impurity I, Carbenicillin Impurity I, Benzylpenicillin Impurity D |
| InChI Formula | InChI=1S/C16H18N2O4S/c1-16(2)12(15(21)22)18-10(8-9-6-4-3-5-7-9)17-11(14(19)20)13(18)23-16/h3-7,11-13H,8H2,1-2H3,(H,19,20)(H,21,22)/t11-,12-,13+/m0/s1 |
| SMILES | CC1(C)S[C@@H]2[C@H](N=C(Cc3ccccc3)N2[C@H]1C(=O)O)C(=O)O |
| IUPAC Name | (3S,7R,7aR)-5-benzyl-2,2-dimethyl-7,7a-dihydro-3H-imidazo[5,1-b][1,3]thiazole-3,7-dicarboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?