missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3S,5S)-3,5-Bis[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1-hydroxy-cyclohexanecarboxylic Acid Methyl Ester, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | (3S,5S)-3,5-Bis(((1,1-dimethylethyl)dimethylsilyl)oxy)-1-hydroxy-cyclohexanecarboxylic Acid Methyl Ester |
| CAS | 139356-33-5 |
| Molecular Formula | C20H42O5Si2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C20H42O5Si2/c1-18(2,3)26(8,9)24-15-12-16(25-27(10,11)19(4,5)6)14-20(22,13-15)17(21)23-7/h15-16,22H,12-14H2,1-11H3/t15-,16-/m0/s1 |
| SMILES | COC(=O)C1(O)C[C@H](C[C@@H](C1)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C |
| IUPAC Name | methyl (3S,5S)-3,5-bis[[tert-butyl(dimethyl)silyl]oxy]-1-hydroxycyclohexane-1-carboxylate |
| Molecular Weight (g/mol) | 418.72 |
| Formula Weight | 418.2571 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction