missing translation for 'onlineSavingsMsg'
Learn More

(3S,5R,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid Calcium Salt, TRC

Product Code. 30354880
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30354880 10 mg -
30351670 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30354880 Supplier Toronto Research Chemicals Supplier No. TRCC98861010MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254452-88-5
Molecular Formula C25H24FNO4 .1/2Ca
Molecular Weight (g/mol) 440.49
Synonym ent-NK-104, Pitavastatin 3S,5R-Isomer Calcium Salt
IUPAC Name (E,3S,5R)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoic acid
SMILES [Ca+2].O[C@@H](C[C@@H](O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4)CC(=O)[O-].O[C@@H](C[C@@H](O)\C=C\c5c(nc6ccccc6c5c7ccc(F)cc7)C8CC8)CC(=O)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C25H24FNO4/c26-17-9-7-15(8-10-17)24-20-3-1-2-4-22(20)27-25(16-5-6-16)21(24)12-11-18(28)13-19(29)14-23(30)31/h1-4,7-12,16,18-19,28-29H,5-6,13-14H2,(H,30,31)/b12-11+/t18-,19-/m0/s1
Formula Weight 880.2848
Chemical Name or Material (3S,5R,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic Acid Calcium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.