missing translation for 'onlineSavingsMsg'
Learn More

(3S,4S)-3-Hexyl-4-((S)-2-hydroxypentadecyl)oxetan-2-one, TRC

Product Code. 30401429
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401429 2.5 mg 2.50mg
30396498 10 mg 10mg
30403946 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401429 Supplier Toronto Research Chemicals Supplier No. TRCH2965332.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3S,4S)-3-Hexyl-4-((S)-2-hydroxypentadecyl)oxetan-2-one
Analyte or Component Names (3S,4S)-3-Hexyl-4-((S)-2-hydroxypentadecyl)oxetan-2-one
Shipping Condition Room Temperature
Molecular Formula C24 H46 O3
Synonym (3S,4S,2'S)-3-Hexyl-4-(2'-hydroxypentadec-1'-yl) Oxetan-2-one
InChI Formula InChI=1S/C24H46O3/c1-3-5-7-9-10-11-12-13-14-15-16-18-21(25)20-23-22(24(26)27-23)19-17-8-6-4-2/h21-23,25H,3-20H2,1-2H3/t21-,22-,23-/m0/s1
Solution Type Neat
SMILES CCCCCCCCCCCCC[C@H](O)C[C@@H]1OC(=O)[C@H]1CCCCCC
Industry Type Pharmaceutical
Molecular Weight (g/mol) 382.62
Quantity 2.5 mg
Formula Weight 382.345
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.