missing translation for 'onlineSavingsMsg'
Learn More

(3S)-3-[4-[(5-Bromo-2-chlorophenyl)methyl]phenoxy]tetrahydrofuran, TRC

Product Code. 30138414
Change view
Click to view available options
Quantity:
1 g
25 g
50 g
Unit Size:
1g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138414 25 g 25g
30137526 50 g 50g
17913815 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138414 Supplier Toronto Research Chemicals Supplier No. TRCB68075525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 915095-89-5
Molecular Formula C17H16BrClO2
Molecular Weight (g/mol) 367.66
Synonym Furan, 3-[4-[(5-bromo-2-chlorophenyl)methyl]phenoxy]tetrahydro-, (3S)- (9CI, ACI), (3S)-3-[4-[(5-Bromo-2-chlorophenyl)methyl]phenoxy]tetrahydrofuran (ACI), (3S)-3-[4-[(5-Bromo-2-chlorophenyl)methyl]phenoxy]oxolane, (S)-3-(4-(5-Bromo-2-chlorobenzyl)phenoxy)tetrahydrofuran, (S)-4-Bromo-1-chloro-2-(4-tetrahydrofuran-3-yloxy-benzyl)-benzene, Empagliflozin Impurity I, (S)-3-(4-(5-Bromo-2-chlorobenzyl)phenoxy)tetrahydrofuran;
IUPAC Name (3S)-3-[4-[(5-bromo-2-chlorophenyl)methyl]phenoxy]oxolane
SMILES Clc1ccc(Br)cc1Cc2ccc(O[C@H]3CCOC3)cc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C17H16BrClO2/c18-14-3-6-17(19)13(10-14)9-12-1-4-15(5-2-12)21-16-7-8-20-11-16/h1-6,10,16H,7-9,11H2/t16-/m0/s1
Formula Weight 366.0022
Chemical Name or Material (3S)-3-[4-[(5-Bromo-2-chlorophenyl)methyl]phenoxy]tetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.