missing translation for 'onlineSavingsMsg'
Learn More

(3S)-1-Azabicyclo[2.2.2]octan-3-amine Dihydrochloride ((3S)-3-Aminoquinuclidine Dihydrochloride), Mikromol™

Product Code. 30049647
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049647 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049647 Supplier Mikromol Supplier No. MM3779.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3S)-1-Azabicyclo[2.2.2]octan-3-amine Dihydrochloride ((3S)-3-Aminoquinuclidine Dihydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3S)-3-Aminoquinuclidine Dihydrochloride
CAS 119904-90-4
Analyte or Component Names (3S)-3-Aminoquinuclidine Dihydrochloride
Shipping Condition Room Temperature
Molecular Formula C7 H14 N2 . 2 Cl H
Certifications/Compliance ISO 17025
Synonym 1-Azabicyclo[2.2.2]octan-3-amine, hydrochloride (1:2), (3S)-, 1-Azabicyclo[2.2.2]octan-3-amine, dihydrochloride, (3S)- (9CI), 1-Azabicyclo[2.2.2]octan-3-amine, dihydrochloride, (S)-, (3S)-1-Azabicyclo[2.2.2]octan-3-amine dihydrochloride, (S)-(-)-3-Aminoquinuclidine dihydrochloride, (S)-3-Aminoquinuclidine dihydrochloride, (S)-Quinuclidin-3-amine dihydrochloride, 3(S)-Aminoquinuclidine dihydrochloride
InChI Formula InChI=1S/C7H14N2.2ClH/c8-7-5-9-3-1-6(7)2-4-9;;/h6-7H,1-5,8H2;2*1H/t7-;;/m1../s1
SMILES Cl.Cl.N[C@@H]1CN2CCC1CC2
IUPAC Name (3S)-quinuclidin-3-amine;hydrochloride
Molecular Weight (g/mol) 199.121
Quantity 25 mg
Formula Weight 198.0691 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.