missing translation for 'onlineSavingsMsg'
Learn More

(3S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid, Mikromol™

Product Code. 30051259
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
30051259 25 mg 25mg
30049657 100 mg 100mg
30463481 4 x 25 mg -
3 options
This item is not returnable. View return policy

Product Code. 30051259

Brand: Mikromol MM0628.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid
CAS 74163-81-8
Analyte or Component Names (3S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H11 N O2
Certifications/Compliance ISO 17025
Synonym Quinapril Hydrochloride Imp. A (EP), Quinapril Imp. A (EP), (3S)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid, Quinapril Hydrochloride Impurity A, Quinapril Impurity A
InChI Formula InChI=1S/C10H11NO2/c12-10(13)9-5-7-3-1-2-4-8(7)6-11-9/h1-4,9,11H,5-6H2,(H,12,13)/t9-/m0/s1
SMILES OC(=O)[C@@H]1Cc2ccccc2CN1
IUPAC Name (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 177.2
Quantity 25 mg
Formula Weight 177.079 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.